Learn More
CD1b/CD1c Monoclonal Antibody (B-B5), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen AHS0198
Additional Details : Weight : 0.05000kg
Description
This gene encodes a member of the CD1 family of transmembrane glycoproteins, which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells. The human genome contains five CD1 family genes organized in a cluster on chromosome 1. The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands. The protein encoded by this gene localizes to late endosomes and lysosomes via a tyrosine-based motif in the cytoplasmic tail, and requires vesicular acidification to bind lipid antigens.
CD1b/CD1c Monoclonal antibody specifically detects CD1b/CD1c in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
CD1b/CD1c | |
Monoclonal | |
FITC | |
CD1C | |
RP11-101J8.3; RP11-101J8.4 | |
Mouse | |
purified | |
RUO | |
910, 911 | |
4° C, store in dark | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
B-B5 | |
PBS with 5% BSA and 0.01% sodium azide; pH 7.2 | |
P29016, P29017 | |
CD1B, CD1C | |
Thymus cells, Jurkat (T cell line), Jurkat (T cell line) | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |