missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD146 Monoclonal Antibody (P1H12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA125100
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: human tongue. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD146 Monoclonal antibody specifically detects CD146 in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
CD146 | |
Monoclonal | |
1.9 mg/mL | |
0.01M PBS with 15mM sodium azide; pH 7.4 | |
P43121 | |
MCAM | |
Human umbilical cord cells. | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
P1H12 | |
Unconjugated | |
MCAM | |
1-gicerin; AV025631; CD146; CD149; Cell surface glycoprotein MUC18; cell surface glycoprotein P1H12; Gicerin; HEMCAM; l-gicerin; l-gicerin precursor; l-gicerin protein; LOW QUALITY PROTEIN: cell surface glycoprotein MUC18; MCAM; melanoma adhesion molecule; melanoma cell adhesion molecule; Melanoma-associated antigen A32; melanoma-associated antigen MUC18; Muc18; mucin-18; s-endo; S-endo 1 endothelial-associated antigen; s-gicerin; s-gicerin precursor | |
Mouse | |
purified | |
RUO | |
4162 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |