missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OX40L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544953
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: QEVNISLHYQ KDEEPLFQLK KVRSVNSLMV ASLTYKDKVY LNVTTDNTSL Sequence homology: Human: 100%
OX40L Polyclonal antibody specifically detects OX40L in Human samples. It is validated for Western Blot.Specifications
OX40L | |
Polyclonal | |
Unconjugated | |
Tnfsf4 | |
Ath1; Ath-1; atherosclerosis 1; CD134 ligand; CD134L; CD252; glycoprotein Gp34; GP34; OX40 antigen ligand; OX40 ligand; OX40L; OX-40L; OX4OL; TAX transcriptionally-activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 (34kD); tax-transcriptionally activated glycoprotein 1 ligand; TNFSF4; TNLG2B; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor (ligand) superfamily, member 4; tumor necrosis factor ligand 2B; tumor necrosis factor ligand superfamily member 4; tumor necrosis factor superfamily member 4; TXGP1; Txgp1l | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7292 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P23510 | |
Tnfsf4 | |
Synthetic peptide directed towards the middle region of human TNFSF4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |