missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OX40 Monoclonal Antibody (OX-86), FITC, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA517916
Additional Details : Weight : 0.01000kg
Description
OX40 Monoclonal antibody specifically detects OX40 in Mouse samples. It is validated for Flow Cytometry.Specifications
OX40 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P47741 | |
Tnfrsf4 | |
Recombinant mouse OX-40 antigen | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Flow Cytometry | |
OX-86 | |
FITC | |
Tnfrsf4 | |
ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; Tnfrsf4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4; Txgp1; Txgp1l | |
Rat | |
Protein G | |
RUO | |
22163 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |