missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Kit Monoclonal Antibody (104D2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110072
Additional Details : Weight : 0.01000kg
Description
This antibody detects an extracellular part of CD117 / c-Kit protooncogen.
c-Kit Monoclonal antibody specifically detects c-Kit in Bovine, Human, Non-human Primate samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.Specifications
c-Kit | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P10721, P43481 | |
KIT | |
MOLM-1 megakaryocytic cells | |
100 μg | |
Primary | |
Bovine, Human, Non-human Primate | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation | |
104D2 | |
Unconjugated | |
KIT | |
belly-spot; Bs; CD117; ckit; C-Kit; c-KIT gene1; c-kit protein; c-kit proto-oncogene protein; c-kit receptor; c-kit receptor tyrosine kinase; dominant spotting; Dominant white spotting; Fdc; Gsfsco1; Gsfsco5; Gsfsow3; KIT; kit oncogene; KIT proto-oncogene receptor tyrosine kinase; KIT proto-oncogene, receptor tyrosine kinase; mast cell growth factor receptor; mast/stem cell growth factor receptor; mast/stem cell growth factor receptor Kit; MGF; p145 c-kit; PBT; Piebald trait protein; protein kinase; proto-oncogene c-Kit; proto-oncogene tyrosine-protein kinase Kit; receptor tyrosine kinase c-kit; receptor tyrosine kinase type III; SCFR; SCO1; SCO5; Sl; soluble KIT variant 1; SOW3; spotted sterile male; Ssm; Steel Factor Receptor; Tr-kit; tyrosine kinase receptor; tyrosine kinase receptor c-KIT; tyrosine-protein kinase Kit; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein; W | |
Mouse | |
Protein A | |
RUO | |
280832, 3815 | |
4° C, do not freeze | |
Liquid |