missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD109 Monoclonal Antibody (496920), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524208
Additional Details : Weight : 0.00010kg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
CD109 Monoclonal antibody specifically detects CD109 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin).Specifications
CD109 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q6YHK3 | |
CD109 | |
Mouse myeloma cell line NS0-derived recombinant human CD109 Val22-Ser1268 (Tyr703Ser and Thr1241Met) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
496920 | |
Unconjugated | |
CD109 | |
150 kDa TGF-beta-1-binding protein; 9930012E15Rik; activated T-cell marker CD109; AI480638; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7; Cd109; CD109 antigen; CD109 molecule; CPAMD7; GARP; Gov platelet alloantigens; GPI-anchored alpha 2 macroglobulin-related protein; GPI-anchored alpha-2 macroglobulin-related protein; p180; platelet-specific Gov antigen; r150; RP11-525G3.1 | |
Mouse | |
Protein A/G | |
RUO | |
135228 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |