missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD105 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529554
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: K562. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD105 Polyclonal antibody specifically detects CD105 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CD105 | |
Polyclonal | |
Unconjugated | |
ENG | |
AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2022 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P17813 | |
ENG | |
Recombinant fragment corresponding to a region within amino acids 1 and 301 of Human CD105 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |