missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-2 Monoclonal Antibody (mIC2/4 (3C4)), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA516837
Additional Details : Weight : 0.01000kg
Description
This product is a low-endotoxin, preservative-free formulation. Endotoxin level is <0.01 EU/ug. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
ICAM-2 Monoclonal antibody specifically detects ICAM-2 in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunoprecipitation.Specifications
ICAM-2 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P35330 | |
ICAM2 | |
COS cells transfected with murine ICAM-2 | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry, Functional Assay, Immunoprecipitation | |
mIC2/4 (3C4) | |
Unconjugated | |
ICAM2 | |
CD102; Icam2; ICAM-2; Intercellular adhesion molecule 2; Ly-60; Lymphocyte function-associated AG-1 counter-receptor; RP23-418O11.10-001 | |
Rat | |
Protein G | |
RUO | |
15896 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |