Learn More
Btk Monoclonal Antibody (7F12H4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515337
Additional Details : Weight : 0.01000kg
Description
MA5-15337 targets BTK in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human and Non-human primate samples. The MA5-15337 immunogen is purified recombinant fragment of BTK expressed in E. Coli. MA5-15337 detects BTK which has a predicted molecular weight of approximately 77kDa.
Btk Monoclonal antibody specifically detects Btk in Human, Non-human Primate samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
Btk | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q06187 | |
BTK | |
Purified recombinant fragment of BTK expressed in E. Coli. | |
RUO | |
695 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
7F12H4 | |
Unconjugated | |
BTK | |
agammaglobulinaemia tyrosine kinase; agammaglobulinemia tyrosine kinase; AGMX1; AI528679; AT; ATK; B-cell progenitor kinase; BPK; Bruton agammaglobulinemia tyrosine kinase; Bruton tyrosine kinase; Bruton's tyrosine kinase; BTK; dominant-negative kinase-deficient Brutons tyrosine kinase; IMD1; Kinase EMB; MGC126261; MGC126262; OTTHUMP00000023677; PSCTK1; RP1-164F3.2; truncated Bruton agammaglobulinemia tyrosine kinase; tyrosine-protein kinase BTK; Tyrosine-protein kinase BTK (Brutons tyrosine kinase) (Agammaglobulinaemia tyrosine kinase) (ATK) (B cell progenitor kinase) (BPK) (Kinase EMB); tyrosine-protein kinase BTK isoform (lacking exon 13 to 17); tyrosine-protein kinase BTK isoform (lacking exon 14); xid; XLA; X-linked immune deficiency | |
Mouse | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |