missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540825
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: LRSQDLDEEL GSTAAGEIVE ADVAIGKGDE TPLTNVKTEA SPESMLSPSH Sequence homology: Cow: 92%; Dog: 92%; Horse: 92%; Human: 100%; Rabbit: 85%
BRD8 Polyclonal antibody specifically detects BRD8 in Human samples. It is validated for Western Blot.Specifications
BRD8 | |
Polyclonal | |
Unconjugated | |
BRD8 | |
2610007E11Rik; 4432404P07Rik; BRD8; bromodomain containing 8; bromodomain-containing protein 8; LOW QUALITY PROTEIN: bromodomain-containing protein 8; p120; Skeletal muscle abundant protein; Skeletal muscle abundant protein 2; SMAP; SMAP2; Thyroid hormone receptor coactivating protein of 120 kDa; trCP120 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10902 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H0E9 | |
BRD8 | |
Synthetic peptide directed towards the middle region of human BRD8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |