missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530491
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, Jurakt, Raji. Predicted reactivity: Mouse (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BRD1 Polyclonal antibody specifically detects BRD1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
BRD1 | |
Polyclonal | |
Unconjugated | |
BRD1 | |
1110059H06Rik; AI316859; BR140-like protein; BRD1; BRL; Bromodomain and PHD finger-containing protein 2; bromodomain containing 1; bromodomain-containing protein 1; BRPF1; BRPF2; DKFZp686F0325; mKIAA4191; RP3-522J7.4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23774 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.94 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O95696 | |
BRD1 | |
Recombinant protein fragment corresponding to a region within amino acids 574 and 878 of Human BRD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |