missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533641
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Gibbon, Monkey, Marmoset (100%) Elephant, Rabbit (94%).
GPR15 Polyclonal antibody specifically detects GPR15 in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Specifications
GPR15 | |
Polyclonal | |
Unconjugated | |
GPR15 | |
4933439K08Rik; BoB; brother of Bonzo; G protein-coupled receptor 15; GPR15; Gpr-15; G-protein coupled receptor 15; MGC126828; MGC126830 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2838 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P49685 | |
GPR15 | |
Synthetic 16 amino acid peptide from 1st cytoplasmic domain of human GPR15. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |