Learn More
BLZF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530714
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Huh7, Hep3B, HepG2, NIH-3T3, BLZF-transfected 293T. Predicted reactivity: Mouse (84%), Rat (84%), Chimpanzee (99%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BLZF1 Polyclonal antibody specifically detects BLZF1 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
BLZF1 | |
Polyclonal | |
Unconjugated | |
BLZF1 | |
1700030G05Rik; Basic leucine zipper nuclear factor 1; basic leucine zipper nuclear factor 1 (JEM-1); Blzf1; blzf1 protein; Blzf1l; cytoplasmic protein; fe06a04; Golgin45; Golgin-45; Golgin-45; golgin-45; JEM1; JEM-1; JEM-1s; JEM-1short protein; LOC504820 protein; MGC22497; p45 basic leucine-zipper nuclear factor; wu:fe06a04; zgc:66280; zgc:85871 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
66352, 8548 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8R2X8, Q9H2G9 | |
BLZF1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 284 of Human BLZF1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |