Learn More
BCAS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530373
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Neuro2A, C8D30, NIH-3T3, Raw264.7, C2C12, PC-12, Rat-2, NT2D1, IMR32, MCF-7. Predicted reactivity: Mouse (100%), Rat (98%), Zebrafish (82%), Xenopus laevis (87%), Pig (97%), Chicken (95%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BCAS2 Polyclonal antibody specifically detects BCAS2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
BCAS2 | |
Polyclonal | |
Unconjugated | |
BCAS2 | |
6430539P16Rik; AI132645; BCAS2; breast carcinoma amplified sequence 2; breast carcinoma-amplified sequence 2; Breast carcinoma-amplified sequence 2 homolog; C76366; C80030; DAM1; DNA amplified in mammary carcinoma 1 protein; pre-mRNA-splicing factor SPF27; Snt309; SPF27; spliceosome associated protein, amplified in breast cancer; spliceosome-associated protein SPF 27 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10286, 295334, 68183 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O75934, Q9D287 | |
BCAS2 | |
Recombinant protein fragment corresponding to a region within amino acids 19 and 204 of Human BCAS2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |