missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAZ2B Mouse anti-Human, Biotin, Clone:, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 730049
Additional Details : Weight : 0.01000kg
Description
730051 is a biotinylated Fab fragment targeting BRG1.
Specifications
BAZ2B | |
Monoclonal | |
Biotin | |
Q9UIF8 | |
BAZ2B | |
2054-2168 of 2168 of BAZ2B Protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunoprecipitation | |
0.5 mg/mL | |
BAZ2B | |
BAZ2B, bromodomain adjacent to zinc finger domain, 2B, DKFZP434H071, DKFZp762I0516, FLJ45644, WALp4 | |
Mouse | |
Affinity chromatography | |
RUO | |
29994 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |