Learn More
BAG6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531213
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (97%), Rat (96%), Pig (98%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BAG6 Polyclonal antibody specifically detects BAG6 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
BAG6 | |
Polyclonal | |
Unconjugated | |
Bag6 | |
2410045D21Rik; AA408914; BAG 6; BAG family molecular chaperone regulator 6; Bag6; BAG-6; BAT 3; BAT3; BCL2 associated athanogene 6; BCL2-associated athanogene 6; D17H6S52E; D6S52E; DAQB-195H10.3; G3; HLA-B associated transcript 3; HLA-B associated transcript-3; HLA-B-associated transcript 3; Large proline-rich protein BAG6; large proline-rich protein BAT3; protein G3; Protein Scythe; scythe | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7917 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.9 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P46379 | |
Bag6 | |
Recombinant fragment corresponding to a region within amino acids 1 and 214 of Human BAT3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |