Learn More
BACE1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532670
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Marmoset, Mouse, Rat, Dog, Bat, Bovine, Hamster, Elephant, Panda, Horse, Rabbit, Pig, Opossum, Guinea pig (100%); Platypus (87%); Xenopus, Pufferfish, Zebrafish, Stickleback (80%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
BACE1 Polyclonal antibody specifically detects BACE1 in Bat, Bovine, Equine, Guinea Pig, Hamster, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat samples. It is validated for Immunohistochemistry (Paraffin).Specifications
BACE1 | |
Polyclonal | |
Unconjugated | |
BACE1 | |
APP beta-secretase; asp 2; ASP2; Aspartyl protease 2; BACE; BACE 1; BACE1; BACE-1D; BACE-I-432; Beta secretase; Beta-secretase 1; beta-secretase 1 precursor variant 1; beta-site amyloid beta A4 precursor protein-cleaving enzyme; beta-site amyloid precursor protein cleaving enzyme 1; Beta-site APP cleaving enzyme 1; beta-site APP-cleaving enzyme 1; C76936; HSPC104; hypothetical protein LOC403005; KIAA1149; Memapsin; Memapsin2; Memapsin-2; membrane-associated aspartic protease 2; protease; transmembrane aspartic proteinase Asp2; zgc:77409 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100071137, 100135485, 100343442, 100511707, 100762079, 23621, 23821, 29392, 614333 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P56817, P56818, P56819, Q1KLR6, Q2HJ40 | |
BACE1 | |
Synthetic 15 amino acid peptide from internal region of human BACE1. | |
50 μg | |
Primary | |
Bat, Rat, Bovine, Equine, Hamster, Hamster, Mouse, Mouse, Non-human Primate, Rabbit, Rabbit | |
Antibody | |
IgG |