Learn More
Anthrax LF Antibody (5F502.2) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-56592CL1
This item is not returnable.
View return policy
Description
Anthrax LF Monoclonal antibody specifically detects Anthrax LF in Human samples. It is validated for Western Blot
Specifications
| Anthrax LF | |
| Monoclonal | |
| CoraFluor 1 | |
| Anthrax lethal factor, Anthrax lethal toxin endopeptidase component, Anthrax LF, B.anthracis lethal factor, bacillus anthracis lethal factor, Lef, LF | |
| The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 779-792 of the Anthrax LF protein (approx. 90 kD). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Western Blot | |
| 5F502.2 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Human | |
| Purified |