Learn More
Anthrax LF Antibody (5F502.2) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NB100-56592CL1
Product Code. 30172000
  • 9225.00 NOK / 0.10mL
Description
Anthrax LF Monoclonal antibody specifically detects Anthrax LF in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Anthrax LF
Monoclonal
CoraFluor 1
Anthrax lethal factor, Anthrax lethal toxin endopeptidase component, Anthrax LF, B.anthracis lethal factor, bacillus anthracis lethal factor, Lef, LF
The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 779-792 of the Anthrax LF protein (approx. 90 kD).
0.1 mL
Primary
Store at 4°C in the dark. Do not freeze.
IgG1 κ
Western Blot
5F502.2
PBS
Mouse
Protein G purified