Learn More
Tocris Bioscience™ ANR 94
GREENER_CHOICE
Product Code. 18083872
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18083872
Brand: Tocris Bioscience™ 3937/50
Adenosine A2A antagonist
Adenosine A2A receptor (AA2AR) antagonist (Ki values are 643 and 46 nM for rat and human AA2ARs respectively). Most active AA2AR antagonist for human receptors. Displays activity in the treatment of Parkinson's disease in vivo; improves parkinsonian motor deficits and tremors. Exhibits neuroprotective and anti-inflammatory effects.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 634924-89-3
Molecular Formula C9H13N5O
Specifications
Quantity 50 mg
Formula Weight 207.23
Purity >99%
Molecular Weight (g/mol) 207.237
Chemical Name or Material ANR 94