Learn More
Masterflex™ Animal Free Female Hex Luer Fittings
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Cole-Parmer™ Animal Free Female Hex Luer Fittings offer superior chemical resistance and are suited for ETO and gamma sterilization.
Brand:  Masterflex™ 30800-00
Additional Details : Weight : 0.02000kg
Product Code. 15342985
  • 590.00 NOK / Pack of 25
Estimated Shipment: 30-07-2024
Description
  • Animal-free polypropylene female luer
  • Available in packs of 25
  • Not recommended for autoclaving
Specifications
Specifications
Fitting
1/8 in.
Female luer to 1/8 in. fitting
Clear
25 Pack