Learn More
Amydricaine Hydrochloride, LoGiCal
Product Code. 30458325
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. 30458325
Brand: LoGiCal LGCFOR1819.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Amydricaine Hydrochloride
CAS 631-38-9
Analyte or Component Names Amydricaine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C16 H26 N2 O2 . Cl H
Certifications/Compliance ISO 9001
Synonym 2-Butanol, 1-(dimethylamino)-2-[(dimethylamino)methyl]-, benzoate (ester), monohydrochloride (8CI,9CI), 2-Benzoxyl-2-[(dimethylamino)methyl]-1-(dimethylamino)butane hydrochloride, Alipine, Alypin hydrochloride, Amydricaine hydrochloride, 2-Butanol, 1-(dimethylamino)-2-[(dimethylamino)methyl]-, 2-benzoate, hydrochloride (1:1), Amydricaine Hydrochloride
InChI Formula InChI=1S/C16H26N2O2.ClH/c1-6-16(12-17(2)3,13-18(4)5)20-15(19)14-10-8-7-9-11-14;/h7-11H,6,12-13H2,1-5H3;1H
Solution Type Neat
SMILES Cl.CCC(CN(C)C)(CN(C)C)OC(=O)c1ccccc1
IUPAC Name [1-(dimethylamino)-2-[(dimethylamino)methyl]butan-2-yl] benzoate;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 314.85
Quantity 5 mg
Formula Weight 314.1761 g/mol