missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048471
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
7783-20-2
H8N2O4S
132.13
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
6097028
CHEBI:62946
N.N.OS(O)(=O)=O
This item is not returnable. View return policy
7783-20-2
H8N2O4S
132.13
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
6097028
CHEBI:62946
N.N.OS(O)(=O)=O

In biochemistry it is used for purification proteins via ammonium sulfate precipitation method. Ammonium sulfate is used in agriculture as fertilizer; it is a slow releasing nitrogenous fertilizer and a rich source of sulphur nutrient. It is employed as a dough conditioner in bread products. It finds use as raw materials for various chemical products such as dyes and pharmacetical products. It is a component of flame-proofing agents and fire extinguisher powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼280°C (decomposition)
Color White
Physical Form Crystalline
Quantity 10 g
Linear Formula (NH4)2SO4
Merck Index 14,555
Solubility Information Highly soluble in water. It is insoluble in acetone,ether and ethanol. It is also insoluble in liquid ammonia.
IUPAC Name sulfuric acid diamine
Formula Weight 132.14
Percent Purity 99.999%
Grade Puratronic™
Density 1.769 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'