missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachlorostannate(IV), Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7051207
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
16960-53-5
Cl6H8N2Sn
367.488
MFCD00036340
UDMAVAHFQCVSSE-UHFFFAOYSA-J
ammonium tin chloride, stannic ammonium chloride, ammonium hexachlorostannate, diammonium hexachlorostannate, ammonium hexachlorostannate 2-, diammonium hexachlorostannate 2-, ammonium tin 4+ chloride 2:1:6, diazanium tin 4+ hexachloride, stannate 2-, hexachloro-, diammonium, oc-6-11, stannate 2-, hexachloro-, ammonium 1:2 , oc-6-11
167595
diazanium;tin(4+);hexachloride
[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]
This item is not returnable. View return policy
16960-53-5
Cl6H8N2Sn
367.488
MFCD00036340
UDMAVAHFQCVSSE-UHFFFAOYSA-J
ammonium tin chloride, stannic ammonium chloride, ammonium hexachlorostannate, diammonium hexachlorostannate, ammonium hexachlorostannate 2-, diammonium hexachlorostannate 2-, ammonium tin 4+ chloride 2:1:6, diazanium tin 4+ hexachloride, stannate 2-, hexachloro-, diammonium, oc-6-11, stannate 2-, hexachloro-, ammonium 1:2 , oc-6-11
167595
diazanium;tin(4+);hexachloride
[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 10 g
Linear Formula (NH4)2SnCl6
Solubility Information Soluble in water
Formula Weight 367.48
Percent Purity 99.999%
Grade Puratronic™
Density 2.4 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium hexachlorostannate(IV)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'