missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium chloride, 99.5% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2 kg
Description
It act as a buffer, as a flux in galvanizing steel sheet. It is used in batteries, to clean soldering irons, analytical lab reagent, in washing powders, electroplating, lustering cotton.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 340° (sublimation) |
Physical Form | Granules |
Quantity | 500 g |
Assay Percent Range | 99.5% min. |
Solubility Information | Soluble in liquid ammonia,acetone,hydrazine,alcohol. Insoluble in diethyl ether,ethyl acetate. |
IUPAC Name | amine hydrochloride |
Formula Weight | 53.49 |
Odor | Odorless |
Refractive Index | 1.642 |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'