missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amikacin disulfate salt, Thermo Scientific Chemicals
621.45 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 220.0°C to 230.0°C |
pH | 2 to 4 (1% aq. soln.) |
Loss on Drying | 13% max. (110°C, 3 h, 5 mm Hg) |
Infrared Spectrum | Authentic |
Quantity | 5 g |
Solubility Information | Solubility in water: ca 50g/L. |
Formula Weight | 781.76 |
Chemical Name or Material | Amikacin disulfate salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'