Learn More
Invitrogen™ Large Volume Upgrade Starter Kit
Large-volume format consumables allow RNA isolation from 3mL of whole blood
Brand:  Invitrogen™ 4344440
Additional Details : Weight : 0.22680kg
Product Code. 15652222
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Includes: Volume adapter plate, elution vial, cap plugs, reusable tube collection plates, microcentrifuge tubes, RNA filters, and two different sizes of reservoirs.
Description
Description
The Large Volume Upgrade Starter Kit upgrades the ABI PRISM™ 6100 Nucleic Acid PrepStation for sample preparation of large volume Tempus™ Blood RNA Tubes.
DNA and RNA Purification and Analysis, General RNA Purification Reagents and Accessories, RNA Extraction, Total RNA Isolation, Total RNA from Blood
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
6100 Nucleic Acid PrepStation
Volume adapter plate, elution vial, cap plugs, reusable tube collection plates, microcentrifuge tubes, RNA filters, and two different sizes of reservoirs.
1 Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not for use in diagnostic procedures.