missing translation for 'onlineSavingsMsg'
Learn More

Amberlite IRA-402(OH), ion exchange resin, Thermo Scientific™

Catalog Number p-7024031
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
52439-77-7
MFCD00145564
This item is not returnable. View return policy
52439-77-7
MFCD00145564

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Amberlite IRA-402(OH) ion exchange resin is a strongly basic gel-type anion exchange resin. It is used to adsorb uranium(VI) from sulphate, removal of cyanide and zinc cyanide. It is also used to produce perchlorate and the free nucleoside. Further, it is used for water conditioning.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and acids.
TRUSTED_SUSTAINABILITY
Density 0.656
Merck Index 14,382
Quantity 100 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlite IRA-402(OH), ion exchange resin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'