Learn More
Amberlite™ IRC120 Na ion exchange resin, Thermo Scientific Chemicals
272.41 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Amberlite™ IR-120(Na) ion exchange resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Amberlite™ IR-120(Na) ion exchange resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.
Solubility
Insoluble in water.
Notes
Water softening, dealkalinization, deionization
Amberlite™ IR-120(Na) ion exchange resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.
Solubility
Insoluble in water.
Notes
Water softening, dealkalinization, deionization
Specifications
Merck Index | 14,382 |
Quantity | 100 g |
Solubility Information | Insoluble in water. |
Chemical Name or Material | Amberlite™ IR-120(Na) ion exchange resin |