Learn More
Amberlite™ CG-50 - type 1, ion-exchange resin, Thermo Scientific Chemicals
525.02 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
Specifications
Packaging | Plastic bottle |
Fieser | 01,511; 04,266; 05,355; 06,302 |
Merck Index | 14, 382 |
Quantity | 25 g |
Chemical Name or Material | Amberlite™ CG-50 - type 1 |