missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, plasma standard solution, Specpure™ Al 10,000μg/mL, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Description
Aluminum, plasma standard solution is used as a single-element standard solution for plasma emission spectrometry. It is also used as a standard solution in analytical chemistry and atomic absorption spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Concentration | Al in 5% HCl |
Quantity | 50 mL |
UN Number | UN1789 |
Grade | Specpure™ |
Solubility | Miscible with water |
Physical Form | Liquid |
Chemical Name or Material | Aluminum plasma standard solution, Al 10000μg/mL |
Safety and Handling
Product Identifier
- Aluminum, plasma standard solution, Specpure Al 10,000µg/ml
- Danger
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'