Learn More
Aluminum, plasma standard solution, Specpure™ Al 1000μg/mL, Thermo Scientific Chemicals
Catalog Number p-4824596
Click to view available options
Quantity:
50 mL
100 mL
500 mL
7429-90-5, 7732-18-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
CHEBI:33629
[Al]
7429-90-5, 7732-18-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
CHEBI:33629
[Al]
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Quantity 50 mL
IUPAC Name aluminium
Concentration or Composition (by Analyte or Components) Hydrochloric acid: 5.0%; Aluminum: 0.1%; Water: 94.9%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Chemical Name or Material Aluminum
compliance-icons
Product Identifier
  • Aluminum, plasma standard solution, Specpure Al 1000µg/ml
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid