Learn More
Aluminum Magnesium gauze , alloy 5056, 16 mesh woven from 0.24mm (0.0095 in.) dia. wire, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 046714.GT
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Aluminum Magnesium gauze, Alloy 5056 | |
Al:Mg; 95:5 wt% | |
150 x 150 mm | |
Odorless |
16 mesh woven from 0.24mm (0.0095 in.) dia. wire | |
MFCD00214039 | |
16 Mesh | |
Wire Cloth |