missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Naphtholbenzein, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
α-Naphtholbenzein is used as a pH indicator. (pH 0.0-0.8/ pH 8.2-10.0).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
α-Naphtholbenzein is used as a pH indicator. (pH 0.0-0.8/ pH 8.2-10.0).
Solubility
Soluble in ethanol, acetic acid and acetone. Insoluble in water.
Notes
Incompatible with strong oxidizing agents, acids, water reactive substances, azo, diazo, hydrazines, metals, nitrides, peroxides, hydroperoxides, epoxides and reducing agents.
α-Naphtholbenzein is used as a pH indicator. (pH 0.0-0.8/ pH 8.2-10.0).
Solubility
Soluble in ethanol, acetic acid and acetone. Insoluble in water.
Notes
Incompatible with strong oxidizing agents, acids, water reactive substances, azo, diazo, hydrazines, metals, nitrides, peroxides, hydroperoxides, epoxides and reducing agents.
Specifications
Melting Point | ∼225°C (decomposition) |
Quantity | 5 g |
Beilstein | 3471575 |
Synonym | p-naphtholbenzein, 1-naphtholbenzein, alpha-naphtholbenzein, .alpha.-naphtholbenzein, 4-4-hydroxynaphthyl phenylmethylene naphthalen-1-one, naphtholbenzein, 1 4h-naphthalenone, 4-4-hydroxy-1-naphthalenyl phenylmethylene, 1 4h-naphthalenone,4-4-hydroxy-1-naphthalenyl phenylmethylene, 1 4h-naphthalenone, 4-.alpha.-4-hydroxy-1-naphthyl benzylidene |
Solubility Information | Soluble in ethanol,acetic acid and acetone. Insoluble in water. |
Formula Weight | 374.44 |
Chemical Name or Material | alpha-Naphtholbenzein |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'