Learn More
alpha-Mangostin, MedChemExpress
Product Code. 16478948
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
Product Code. 16478948
Brand: Medchem Express HYN0328
MedChemExpress alpha-Mangostin (α-Mangostin) is a dietary xanthone with broad biological activities, such as antioxidant, anti-allergic, antiviral, antibacterial, anti-inflammatory and anticancer effects. It is an inhibitor of mutant IDH1 (IDH1-R132H) with a Ki of 2.85 μM.
alpha-Mangostin (α-Mangostin) is a dietary xanthone with broad biological activities, such as antioxidant, anti-allergic, antiviral, antibacterial, anti-inflammatory and anticancer effects. It is an inhibitor of mutant IDH1 (IDH1-R132H) with a Ki of 2.85 μM.
Specifications
Chemical Name or Material alpha-Mangostin
Physical Form Solid
CAS 6147-11-1
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C24H26O6
For Use With (Application) Cancer-programmed cell death
Synonym α-Mangostin
Solubility Information DMSO : 110 mg/mL (267.99 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C1C2=C(OC3=C1C(C/C=C(C)\C)=C(OC)C(O)=C3)C=C(O)C(C/C=C(C)\C)=C2O
Molecular Weight (g/mol) 410.46
Formula Weight 410.46
Percent Purity 99.64%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.