missing translation for 'onlineSavingsMsg'
Learn More

ALDH1A1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1A10A2, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60171-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16847145

  • 1415.13 NOK / 20µL
Estimated Shipment: 09-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ALDH1A1 belongs to the aldehyde dehydrogenases family of proteins. Aldehyde dehydrogenase is the second enzyme of the major oxidative pathway of alcohol metabolism. Two major liver isoforms of this enzyme, cytosolic and mitochondrial, can be distinguished by their electrophoretic mobilities, kinetic properties, and subcellular localizations. Most Caucasians have two major isozymes, while approximately 50% of Orientals have only the cytosolic isozyme, missing the mitochondrial isozyme. A remarkably higher frequency of acute alcohol intoxication among Orientals than among Caucasians could be related to the absence of the mitochondrial isozyme.
Specifications

Specifications

ALDH1A1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P00352, P24549, P51647
ALDH1A1
ALDH1A1 Fusion Protein Ag8551
20 μL
Primary
Mouse, Rat, Porcine, Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
1A10A2
Unconjugated
ALDH1A1
ALDC, ALDH E1, ALDH1, ALDH11, ALDH1A1, ALHDII, PUMB1, RALDH 1, RALDH1, Retinal dehydrogenase 1
Mouse
Protein G
RUO
100517137, 11668, 216, 24188
-20°C
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers