missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALADIN Rabbit anti-Human, Non-human primate, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | ALADIN |
---|---|
Concentration | 0.49 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894164
|
Proteintech
15127-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16884164
|
Proteintech
15127-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the WD-repeat family of regulatory proteins and may be involved in normal development of the peripheral and central nervous system. The encoded protein is part of the nuclear pore complex and is anchored there by NDC1. Defects in this gene are a cause of achalasia-addisonianism-alacrima syndrome (AAAS), also called triple-A syndrome or Allgrove syndrome. Two transcript variants encoding different isoforms have been found for this gene.Specifications
ALADIN | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Non-human Primate | |
Q9NRG9 | |
8086 | |
ALADIN Fusion Protein Ag7211 | |
Primary | |
-20°C | |
AAAS |
0.49 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AAA, AAAS, AAASb, ADRACALA, ADRACALIN, ALADIN, GL003 | |
AAAS | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |