missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Al-23 Tube, One End Closed, O.D.: 24mm, I.D.: 18mm, Alfa Aesar™
Brand: Alfa Aesar 030078.EM
Additional Details : Weight : 0.01000kg
Specifications
Al-23 Tube | |
800mm | |
One end closed |