missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT2 (PKB beta) Recombinant Human Protein
AKT2 (PKB beta)
Brand: Invitrogen PV3184
Additional Details : Weight : 3.36000kg
Description
AKT2 is a phosphoinositide-dependent serine⁄threonine protein kinase that is enriched in insulin responsive tissues and has been implicated in the metabolic actions of the hormone.Specifications
NP_001617 | |
Cell Viability, Pharma and Biopharma, Cell Proliferation and Function, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
59.1kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
AKT2 (PKB beta) | |
AKT2 (PKB beta) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM Na3VO4, 1mM DTT, 0.01% Triton™ X-100, 200μM ATP, 50μM crosstide peptide substrate (GRPRTSSFAEG) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
AKT2 (PKB beta) Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.44mg/mL | |
208 | |
AKT2 (PKB beta) Recombinant Human Protein | |
–80°C | |
1 Each | |
AKT2 (PKB beta) Recombinant Human Protein | |
AKT | |
240nmoles of phosphate transferred to Crosstide peptide substrate (GRPRTSSFAEG) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL | |
His Tag | |
Kinase and inhibitors | |
90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.