Learn More
Ajuba Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28686-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The Ajuba antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Ajuba. This antibody recognizes human antigen. The Ajuba antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Ajuba | |
Polyclonal | |
Unconjugated | |
20 μL | |
84962 | |
IgG |
Western Blot, ELISA | |
400 μg/mL | |
Rabbit | |
Primary | |
Human |