missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Airwing B-WR Safety Helmet

A lightweight helmet with an attractive design.

Brand:  Uvex™ 9762030

2596.67 NOK valid until 2023-05-26
Use promo code "20831" to get your promotional price.



Additional Details : Weight : 11.00000kg

 View more versions of this product

Product Code. 17564086

  • 3895.00 NOK / Pack of 25

Estimated Shipment: 17-07-2023
to see stock.
Add to basket

Description

Description

  • Lightweight safety helmet with three air vents, one ventilation zone on the forehead and two at the back
  • 6-point textile suspension harness
  • Side slots for attaching ear defenders or visors
  • Rotating size adjusters
Specifications

Specifications

EN 397
HDPE
Industrial Safety Helmet
Long Brim
White
9762
52 to 61 cm
Yes
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers