missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ AGGF1 Recombinant Protein
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH02828 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 55109 |
Molecular Weight (g/mol) | 37.73 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16131123
|
Abnova™
H00055109-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16141123
|
Abnova™
H00055109-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH02828 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
37.73 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
FLJ10283/GPATC7/GPATCH7/HSU84971/HUS84971/VG5Q | |
AGGF1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
55109 | |
AGGF1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AGGF1 | |
Human | |
Recombinant | |
Solution |