Learn More
Hettich™ Adapter for Hettich Rotors, for Carrier no. 1366
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 1326
Additional Details : Weight : 0.80000kg
Product Code. 12043864
  • 850.00 NOK / Each
Estimated Shipment: 17-06-2024
to see stock.
Description
Description
  • Rotor adapter for carrier no. 1366
  • for 12 x 4mL tubes, 12 x 60mm (dia. x L)
  • For use with Universal 320/320 R Centrifuges
  • Universal 320/320 R Centrifuges feature excellent performance and a comprehensive range of accessories
  • The Universal 320/320 R can be used for both standard and special applications, they are not only costeffective, they also save valuable space in the laboratory
Specifications
Specifications
Rotor Adapter
Carrier no. 1366