Learn More
Hettich™ Adapter for Carrier no. 4780
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 4769
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10038142
  • 1985.00 NOK / Each
Estimated Shipment: 19-08-2022
Description
  • Adapter for Carrier no. 4780
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
Rotor Adapter
13 x 15 mL
120 mm
13
Conical
17 mm
Carrier no. 4780