missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACKR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | ACKR2 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888423
|
Proteintech
11386-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16878423
|
Proteintech
11386-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a beta chemokine receptor, which is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptor-mediated signal transduction are critical for the recruitment of effector immune cells to the inflammation site. This gene is expressed in a range of tissues and hemopoietic cells. The expression of this receptor in lymphatic endothelial cells and overexpression in vascular tumors suggested its function in chemokine-driven recirculation of leukocytes and possible chemokine effects on the development and growth of vascular tumors. This receptor appears to bind the majority of beta-chemokine family members; however, its specific function remains unknown. This gene is mapped to chromosome 3p21.3, a region that includes a cluster of chemokine receptor genes.Specifications
ACKR2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
O00590, O08707, O09027 | |
1238, 140473, 59289 | |
ACKR2 Fusion Protein Ag1941 | |
Primary | |
-20°C | |
ACKR2 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ACKR2, Atypical chemokine receptor 2, C C chemokine receptor D6, CCBP2, CCR10, CCR9, chemokine binding protein 2, Chemokine binding protein D6, Chemokine receptor CCR 10, Chemokine receptor CCR 9, CMKBR9, D6, hD6 | |
Ackr2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |