missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Scharlab

Catalog Number 11369773
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in solvents, analytical chemistry, synthesis of organic products, and photography.

  • Synonyms: Dimethyl ketone, 2-Propanone
  • Molecular Formula: C3H6O
  • Molar Mass: 58.08g/mol
  • CAS: 67-64-1
  • EIN ECS No.: 200-662-2
  • Dielectric Constant: 20.7 at 25°C
  • Reducing Substances: Passes test
CAS 67-64-1
Color Colorless
Quantity 1 L
Physical Form Liquid
Molecular Formula C3H6O
MDL Number MFCD00008765
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
PubChem CID 180
ChEBI CHEBI:15347
Formula Weight 58.08g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone, analytical grade, ACS, Reag. Ph Eur
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'