missing translation for 'onlineSavingsMsg'
Learn More

Acetobromo-alpha-D-glucuronic acid methyl ester, 98%, Thermo Scientific Chemicals

Catalog Number p-4228286
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
21085-72-3
C13H17BrO9
397.17
GWTNLHGTLIBHHZ-SVNGYHJRSA-N
2r,3r,4s,5s,6s-2-bromo-6-methoxycarbonyl tetrahydro-2h-pyran-3,4,5-triyl triacetate, acetobromo-alpha-d-glucuronic acid methyl ester, bromomethyltriacetyl glucuronate, bromo-2,3,4-tri-o-acetyl-alpha-d-glucopyranuronic acid methyl ester, methyl acetobromo-alpha-d-glucuronate, 2,3,4-tri-o-acetyl-alpha-d-glucopyranosyl bromide uronic acid methyl ester, methyl 2s,3s,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxane-2-carboxylate, zlchem 1315, acetobromo-alpha-d-glucuronic acid, methyl ester, methyl, a-acetobromoglucuronate
88785
methyl (2S,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxane-2-carboxylate
CC(=O)OC1C(C(OC(C1OC(=O)C)Br)C(=O)OC)OC(=O)C
This item is not returnable. View return policy
21085-72-3
C13H17BrO9
397.17
GWTNLHGTLIBHHZ-SVNGYHJRSA-N
2r,3r,4s,5s,6s-2-bromo-6-methoxycarbonyl tetrahydro-2h-pyran-3,4,5-triyl triacetate, acetobromo-alpha-d-glucuronic acid methyl ester, bromomethyltriacetyl glucuronate, bromo-2,3,4-tri-o-acetyl-alpha-d-glucopyranuronic acid methyl ester, methyl acetobromo-alpha-d-glucuronate, 2,3,4-tri-o-acetyl-alpha-d-glucopyranosyl bromide uronic acid methyl ester, methyl 2s,3s,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxane-2-carboxylate, zlchem 1315, acetobromo-alpha-d-glucuronic acid, methyl ester, methyl, a-acetobromoglucuronate
88785
methyl (2S,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxane-2-carboxylate
CC(=O)OC1C(C(OC(C1OC(=O)C)Br)C(=O)OC)OC(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 80°C to 110°C
Color White to Gray
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation +185° to +195° (20°C, 589nm) (c=1, ethyl acetate)
Stabilizer 2% (CaCO3)
Formula Weight 397.17
Percent Purity 98%
Physical Form Crystalline Powder or Solid
Chemical Name or Material Acetobromo-α-D-glucuronic acid methyl ester
Show More Show Less
compliance-icons
Product Identifier
  • Acetobromo-alpha-D-glucuronic acid methyl ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'