missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ 5.8 mm Connector and 0.2 μm Syringe Disk Filter
Click to view available options
For use with Screw Cap GL 45 with 2-hose connector
Description
- Accessory for use with DURAN™ Screw Cap GL 45 with 2-hose connector (No. 293102807)
- Consists of: Syringe Filter (0.2 μm) with connector, non-sterile 0.2 μm filter and female Luer Slip to 5.8mm male connector, with O-ring seal
Specifications
For Use With (Equipment) | For use with DURAN Screw Cap GL 45 with 2-hose connector (No. 293102807) |
Description | Syringe Filter (0.2 μm) with connector, non-sterile 0.2 μm filter and female Luer Slip to 5.8mm male connector, with O-ring seal |
For Use With (Application) | Sterile fluid transfer in and out of lab bottles; Gas transfer in and out of lab bottles; Fluid transfer bewteen lab bottles; Supply of reagents to instruments |
Material | Polypropylene, EPDM |
Quantity | 2/Pk. |
Autoclavable | Autoclavable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'