missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG1 Antibody [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB400-132JF525
This item is not returnable.
View return policy
Description
ABCG1 Polyclonal antibody specifically detects ABCG1 in Chinese Hamster,Hamster,Human,Monkey,Mouse,Rabbit,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunoblot,In vitro Assay,In vivo Assay,Flow (Intracellular),Immunocytochemistry/ Immunofluorescence
Specifications
| ABCG1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Chinese Hamster, Hamster, Human, Monkey, Mouse, Rabbit, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunoblot, In vitro Assay, In vivo Assay, Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 525 | |
| ABC transporter 8, ABC8, ABCG1, ATP-binding cassette sub-family G member 1, ATP-binding cassette transporter 8, ATP-binding cassette transporter member 1 of subfamily G, ATP-binding cassette, sub-family G (WHITE), member 1, homolog of Drosophila white, MGC34313, White protein homolog, white protein homolog (ATP-binding cassette transporter 8), WHITE1, WHT1 | |
| A synthetic peptide made to an internal region of human ABCG1 (between residues 300-400). [UniProt# P45844] | |
| 0.1 mL | |
| ABC Transporters, Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Neuroscience, Signal Transduction | |
| 9619 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction