missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCA1 Antibody (768510) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7207JF525
This item is not returnable.
View return policy
Description
ABCA1 Monoclonal antibody specifically detects ABCA1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| ABCA1 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1 | |
| E. coli-derived recombinant human ABCA1, Phe1248-Gln1350, Accession # O95477 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 768510 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| ABC Transporters, Astrocyte Markers, Cancer, Cardiovascular Biology, Cellular Markers, Cholesterol Metabolism, Diabetes Research, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Signal Transduction | |
| 19 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction