Learn More
Sino Biological™ AARS Recombinant Mouse Protein, His tag
Alanyl-Trna Synthetase (AARS), recombinant mouse protein is supplied in frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell bio
Brand: Sino Biological™ 50384M08B25
Additional Details : Weight : 0.01000kg
Description
Alanyl-Trna Synthetase (AARS), recombinant mouse protein is supplied in frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.This recombinant protein was expressed from a DNA sequence encoding the mouse AARS (Q8BGQ7) (Met 1-Asn 968) fused to a polyhistidine tag at the C-terminus.
Activity: The activity of this protein has not been tested.
Reconstitution: No reconstitution is needed. Store aliquots under sterile conditions at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles.
Formulation: 20 mM Tris, 500 mM NaCl, 10% Glycerol, pH 7.4.
Expiration Date: Expires one year from date of receipt when stored as instructed.
This protein is manufactured by Sino Biological Inc.
Specifications
Assay Standard | |
108.3 kDa | |
PAGE | |
Store at -80 °C | |
AI316495, C76919, MGC37368, sti | |
AARS | |
Recombinant | |
Insect | |
Frozen |
234734 | |
alanyl-tRNA synthetasep | |
5 x 5μg | |
< 1 EU/μg | |
Includes 5 vials of protein in suspension buffer | |
Mouse | |
His Tag | |
Other Proteins | |
≥ 85% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.